Learn More
American Research Products Inc Monkey C C motif chemokine 17
Catalog No. 5000327429
Encompass
Monkey C-C motif chemokine 17 (CCL17) ELISA kit from CUSABIO Detection Range 39 pg/mL-2500 pg/mL Sensitivity 9 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27429 Supplier American Research Products Inc Supplier No. CSBEL004780RH
Monkey C-C motif chemokine 17 (CCL17) ELISA kit from CUSABIO Detection Range 39 pg/mL-2500 pg/mL Sensitivity 9 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich