Learn More
Life Diagnostics Inc MONKEY C REA PROT (CRP)ELISA
SDP
Supplier:  Life Diagnostics Inc CRP3
Encompass_Preferred
Monkey C-Reactive Protein(CRP) Elisa Test Kit; CRP Range from 0 to 8.3g/mL 1,2; Kit Includes Anti-monkey CRP Antibody, Enzyme Conjugate Reagent, Reference Standard, 10x Diluent, 20x Wash Solution, TMB Reagent, Stop Solution
Catalog No. NC9495745