Learn More
American Research Products Inc Monkey C X C motif chemokine 1
Catalog No. 5000328007
Encompass
Monkey C-X-C motif chemokine 10 (CXCL10) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28007 Supplier American Research Products Inc Supplier No. CSBEL006240RH
Monkey C-X-C motif chemokine 10 (CXCL10) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich