Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Monkey C3a ELISA Kit

Catalog No. EEL212
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL212 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL212 Supplier Invitrogen™ Supplier No. EEL212
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Monkey C3a ELISA Kit quantifies Complement C3a in serum and plasma. The Monkey C3a solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.16-10 ng/mL
Assay Sensitivity 0.09 ng/mL
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum
Target Species Monkey
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3r1; Complement C3; complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3 alpha-chain; complement component C3a; complement component C3b; CPAMD1; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; Plp; prepro-C3Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3r1; Complement C3; complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3 alpha-chain; complement component C3a; complement component C3b; CPAMD1; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; Plp; prepro-C3;
Interassay CV <10%
Intraassay CV <10%
Kit Contents Micro ELISA Plate (Dismountable), Reference Standard , Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis, Reference Standard, Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma: 100 μL, Serum: 100 μL
Shipping Condition Wet Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Complement C3a
Test Time 1 hr 20 min
Total Assay Time 3 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.