Learn More
Kingfisher Biotech Inc Bovine IL-8 ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1263Y004
Encompass_Preferred
The Bovine IL-8 ELISA contains capture antibody, standard, and detection antibody for development of a Bovine IL-8 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Bovine IL-8 ELISA can also be used to measure Zebu IL-8. Alternate names include CXCL8, chemokine (C-X-C motif) ligand 8, GCP-1, GCP1, LECT, LUCT, LYNAP, MDNCF, MONAP, NAF, NAP-1, NAP1, IL8, C-X-C motif chemokine ligand 8, Interleukin-8. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5394