Learn More
American Research Products Inc Monkey circulating immune comp
Catalog No. 5000326786
Encompass
Monkey circulating immune compleX (CIC) ELISA Kit from CUSABIO Detection Range 0 312 g/mL-20 g/mL Sensitivity 0 078 g/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-26786 Supplier American Research Products Inc Supplier No. CSBE15098MK
Monkey circulating immune compleX (CIC) ELISA Kit from CUSABIO Detection Range 0 312 g/mL-20 g/mL Sensitivity 0 078 g/mL Sample serum plasma Method Sandwich