Learn More
Kingfisher Biotech Inc Cynomolgus Monkey CXCL10 Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1181Y005
Encompass
The Cynomolgus Monkey CXCL10 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey CXCL10 applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey CXCL10 yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey CXCL10 Specifications: (Molecular Weight: 8.7 kDa) (Amino Acid Sequence: IPLSRTVRCT CISISNQPVN PRSLEKLEII PPSQFCPHVE IIATMKKKGE KRCLNPESKA IKNLLKAVSK ERSKRSP (77)) (Gene ID: 102134351). For research use only. Made in the USA
Catalog No. 50-253-3584