Learn More
Kingfisher Biotech Inc Monkey CXCL11 Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Catalog No. 502964724
Encompass
The Cynomolgus Monkey CXCL11 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey CXCL11 applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey CXCL11 yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey CXCL11 Specifications: (Molecular Weight:7.5 kDa) (Amino Acid Sequence:GRCLCIGPGV KAVKVADIEK ASIIYPSNNC DKIEVIITLK ENKGQRCLNP KSKQARLIIK KVERKNF). For research use only. Made in the USA
Catalog No. 50-296-4724 Supplier Kingfisher Biotech Inc Supplier No. RP2231Y100
The Cynomolgus Monkey CXCL11 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey CXCL11 applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey CXCL11 yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey CXCL11 Specifications: (Molecular Weight:7.5 kDa) (Amino Acid Sequence:GRCLCIGPGV KAVKVADIEK ASIIYPSNNC DKIEVIITLK ENKGQRCLNP KSKQARLIIK KVERKNF). For research use only. Made in the USA