Learn More
Innovative Research Inc Monkey Cynomolgus IgG Affinity Purified 10mg
Supplier:  Innovative Research Inc IMNCYIGGAP10MG
Encompass_Preferred
Monkey Cynomolgus IgG Affinity Purified from Innovative Research has been purified from normal Cynomolgus Monkey Serum using Protein A affinity chromatography. This is a frozen liquid buffered in 0.02M Sodium Phosphate; 0.15M NaCl; pH 7.4, with a concentration of 18.2 mg/ml and a purity of >95%.
Catalog No. 50-203-6447