Learn More
American Research Products Inc Monkey fibrinogen degradation
Catalog No. 5000328288
Encompass
Monkey fibrinogen degradation product (FDP) ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 312 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28288 Supplier American Research Products Inc Supplier No. CSBEQ027418MK
Monkey fibrinogen degradation product (FDP) ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 312 ng/mL Sample serum plasma tissue homogenates Method Competitive