Learn More
American Research Products Inc Monkey follicle stimulating ho
Catalog No. 5000326794
Encompass
Monkey follicle-stimulating hormone FSH ELISA Kit from CUSABIO Detection Range 0 2 mIU/mL-80 mIU/mL Sensitivity 0 2 mIU/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-26794 Supplier American Research Products Inc Supplier No. CSBE06866MO
Monkey follicle-stimulating hormone FSH ELISA Kit from CUSABIO Detection Range 0 2 mIU/mL-80 mIU/mL Sensitivity 0 2 mIU/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive