Learn More
Kingfisher Biotech Inc Cynomolgus Monkey GM-CSF Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Catalog No. 502533568
Encompass
The Cynomolgus Monkey GM-CSF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey GM-CSF applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey GM-CSF yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey GM-CSF Specifications: (Molecular Weight: 14.4 kDa) (Amino Acid Sequence: APARSPSPGT QPWEHVNAIQ EARRLLNLSR DTAAEMNKTV EVVSEMFDLQ EPSCLQTRLE LYKQGLQGSL TKLKGPLTMM ASHYKQHCPP TPETSCATQI ITFQSFKENL KDFLLVIPFD CWEPVQE (127)) (Gene ID: 102118747). For research use only. Made in the USA
Catalog No. 50-253-3568 Supplier Kingfisher Biotech Inc Supplier No. RP1185Y005
The Cynomolgus Monkey GM-CSF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey GM-CSF applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey GM-CSF yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey GM-CSF Specifications: (Molecular Weight: 14.4 kDa) (Amino Acid Sequence: APARSPSPGT QPWEHVNAIQ EARRLLNLSR DTAAEMNKTV EVVSEMFDLQ EPSCLQTRLE LYKQGLQGSL TKLKGPLTMM ASHYKQHCPP TPETSCATQI ITFQSFKENL KDFLLVIPFD CWEPVQE (127)) (Gene ID: 102118747). For research use only. Made in the USA