Learn More
American Research Products Inc Monkey Hemoglobin subunit beta
Catalog No. 5000327660
Encompass
Monkey Hemoglobin subunit beta (HBB) ELISA kit from CUSABIO Detection Range 0 12 ng/mL-500 ng/mL Sensitivity 0 12 ng/mL Sample lysates for RBC Method Competitive
Catalog No. 50-003-27660 Supplier American Research Products Inc Supplier No. CSBEL010150RH
Monkey Hemoglobin subunit beta (HBB) ELISA kit from CUSABIO Detection Range 0 12 ng/mL-500 ng/mL Sensitivity 0 12 ng/mL Sample lysates for RBC Method Competitive