Learn More
American Research Products Inc Monkey high sensitiVity C Reac
Catalog No. 5000326105
Encompass
Monkey high sensitiVity C-ReactiVe Protein hs-CRP ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26105 Supplier American Research Products Inc Supplier No. CSBE10035MO
Monkey high sensitiVity C-ReactiVe Protein hs-CRP ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich