Learn More
Kingfisher Biotech Inc Cynomolgus Monkey IFN beta Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1709Y025
Encompass
The Cynomolgus Monkey IFN beta yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey IFN beta applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey IFN beta yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey IFN beta Specifications: (Molecular Weight: 20.0 kDa) (Amino Acid Sequence: MSYNLLGFLQ RSSSFQCQKL LWQLNGRLEY CLKDRMNFDI PEEIKQPQQF QKEDAALTIY EMLQNIFAIF RQDLSSTGWN ETIVENLLAN VYHQIDHLKT ILEEKLEKED FTRGKFVSSL HLKRYYGRIL HYLKAKEYSH CAWTIVRVEI LRNFFFINKL TGYLQN (166)) (Gene ID: 3456). For research use only. Made in the USA
Catalog No. 50-253-2913