Learn More
Kingfisher Biotech Inc Cynomolgus Monkey IFN gamma Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1022Y025
Encompass
The Cynomolgus Monkey IFN gamma yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey IFN gamma applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey IFN gamma yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey IFN gamma Specifications: (Molecular Weight: 16.8 kDa) (Amino Acid Sequence: QDPYVKEAEN LKKYFNAGDP DVADNGTLFL DILRNWKEES DRKIMQSQIV SFYFKLFKNF KDDQRIQKSV ETIKEDINVK FFNSNKKKRD DFEKLTNYSV TDSNVQRKAV HELIQVMAEL SPAAKIGKRK RSQMFRGRRA SQ (142)) (Gene ID: 102128291). For research use only. Made in the USA
Catalog No. 50-241-3336