Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Monkey IFN-gamma ELISA Kit

Catalog No. EP8RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EP8RBX5 5 x 96 Tests
EP8RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EP8RBX5 Supplier Invitrogen™ Supplier No. EP8RBX5
Add to Cart
Add to Cart

ELISA

Monkey IFN-gamma quantitates monkey IFN-gamma in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey IFN-gamma.

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01579, P01580, P01581
Assay Range 24-6000 pg/mL
Assay Sensitivity 24 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human, Mouse, Monkey, Rat
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN γ; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFNγ; Immune interferon; Interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon γ; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferonγ; M-IFN-g; R-IFN-gBoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN γ; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFNγ; Immune interferon; Interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon γ; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferonγ; M-IFN-g; R-IFN-g;
Gene ID (Entrez) 15978, 25712, 3458
Gene Symbol IFNG
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target IFN gamma
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.