Learn More
Mabtech Inc Monkey IFN-γ ELISpot Plus Kit, 2 pre-coated plates, Alkaline Phosphatase (ALP) conjugate, Cross-reactive with human IFN-γ, Includes Anti-CD3 mAb
Supplier:  Mabtech Inc 3421M4APW10
Encompass
Monkey IFN-γ ELISpot Plus Kit, 2 pre-coated plates, Alkaline Phosphatase (ALP) conjugate, Cross-reactive with human IFN-γ, Includes Anti-CD3 mAb
Catalog No. NC0872455