Learn More
Life Diagnostics Inc Monkey IgA, 0.1MG
SDP
Catalog No. NC2682395
Encompass_Preferred
Purified from Rhesus or Cynomolgus monkey serum using a combination of affinity and size exclusion chromatography.
Catalog No. NC2682395 Supplier Life Diagnostics Inc Supplier No. 300024
Purified from Rhesus or Cynomolgus monkey serum using a combination of affinity and size exclusion chromatography.