Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Monkey IGF-2 ELISA Kit

Catalog No. EP9RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EP9RBX5 5 x 96 Tests
EP9RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EP9RBX5 Supplier Invitrogen™ Supplier No. EP9RBX5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Monkey IGF-2 quantitates monkey IGF-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey IGF-2.

The IGFs are mitogenic, polypeptide growth factors that stimulate the proliferation and survival of various cell types, including muscle, bone, and cartilage tissue in vitro. IGFs are predominantly produced by the liver, although a variety of tissues produce the IGFs at distinctive times. The IGFs belong to the Insulin gene family, which also contains insulin and relaxin. The IGFs are similar to insulin by structure and function, but have a much higher growth-promoting activity than insulin. IGF-II expression is influenced by placenta lactogen, while IGF-I expression is regulated by growth hormone. Both IGF-I and IGF-II signal through the tyrosine kinase type I receptor (IGF-IR), but IGF-II can also signal through the IGF-II/Mannose-6-phosphate receptor. Mature IGFs are generated by proteolytic processing of inactive precursor proteins, which contain N-terminal and C-terminal propeptide regions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AL033362; C11orf43; FLJ22066; FLJ44734; GRDF; H-IGF-2; IGF; Igf2; Igf-2; IGFII; IGF-II; Insulin like growth factor; insulin like growth factor 2; insulin-like growth factor 2; insulin-like growth factor 2 (somatomedin A); insulin-like growth factor II; insulin-like growth factor II (IGF-II); Insulin-like growth factor II (somatomedin A); Insulin-like growth factor II Ala-25 Del; insulin-like growth factor II isoform 2 variant 4 preprotein; insulin-like growth factor type 2; M6pr; Mpr; MSA; multiplication-stimulating activity; Multiplication-stimulating polypeptide; Peg2; PP1446; PP9974; Preptin; RNIGF2; Somatomedin-A; T3M-11-derived growth factor
Assay Range 0.2-50 ng/mL
Assay Sensitivity 0.2 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Monkey
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AL033362; C11orf43; FLJ22066; FLJ44734; GRDF; H-IGF-2; IGF; Igf2; Igf-2; IGFII; Igf-II; Insulin like growth factor; insulin like growth factor 2; insulin-like growth factor 2; insulin-like growth factor 2 (somatomedin A); insulin-like growth factor II; insulin-like growth factor II (IGF-II); Insulin-like growth factor II (somatomedin A); Insulin-like growth factor II Ala-25 Del; insulin-like growth factor II isoform 2 variant 4 preprotein; insulin-like growth factor type 2; M6pr; Mpr; MSA; multiplication-stimulating activity; Multiplication-stimulating polypeptide; Peg2; PP1446; PP9974; preptin; RNIGF2; Somatomedin-A; T3M-11-derived growth factorAL033362; C11orf43; FLJ22066; FLJ44734; GRDF; H-IGF-2; IGF; Igf2; Igf-2; IGFII; Igf-II; Insulin like growth factor; insulin like growth factor 2; insulin-like growth factor 2; insulin-like growth factor 2 (somatomedin A); insulin-like growth factor II; insulin-like growth factor II (IGF-II); Insulin-like growth factor II (somatomedin A); Insulin-like growth factor II Ala-25 Del; insulin-like growth factor II isoform 2 variant 4 preprotein; insulin-like growth factor type 2; M6pr; Mpr; MSA; multiplication-stimulating activity; Multiplication-stimulating polypeptide; Peg2; PP1446; PP9974; preptin; RNIGF2; Somatomedin-A; T3M-11-derived growth factor;
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target IGF2
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.