Learn More
Invitrogen™ Rhesus Monkey IGFBP-3 ELISA Kit
Description
Monkey IGFBP-3 quantitates monkey IGFBP-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey IGFBP-3.
Specifications
Specifications
| Assay Range | 1.024-250 ng/mL |
| Assay Sensitivity | 1.02 ng/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Monkey |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | acid stable subunit of the 140 K IGF complex; AI649005; binding protein 29; binding protein 53; BP53; BP-53; Growth hormone dependent binding protein; growth hormone-dependent binding protein; I79_011450; IBP3; IBP-3; IGF binding protein 3; IGF-binding protein 3; IGFBP; Igfbp3; IGF-BP3; IGFBP-3; IGgfbp3; Insulin like growth factor binding protein; insulin like growth factor binding protein 3; insulin-like growth factor binding protein 3; Insulin-like growth factor binding protein 3 precursor; insulin-like growth factor-binding protein (IGF-BP3); insulin-like growth factor-binding protein 3; tcag7.703acid stable subunit of the 140 K IGF complex; AI649005; binding protein 29; binding protein 53; BP53; BP-53; Growth hormone dependent binding protein; growth hormone-dependent binding protein; I79_011450; IBP3; IBP-3; IGF binding protein 3; IGF-binding protein 3; IGFBP; Igfbp3; IGF-BP3; IGFBP-3; IGgfbp3; Insulin like growth factor binding protein; insulin like growth factor binding protein 3; insulin-like growth factor binding protein 3; Insulin-like growth factor binding protein 3 precursor; insulin-like growth factor-binding protein (IGF-BP3); insulin-like growth factor-binding protein 3; tcag7.703; |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.