Learn More
Abcam Monkey IgG ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1905491X96TESTS
Encompass_Preferred
Monkey IgG ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Monkey IgG in Monkey in Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1748221