Learn More
Immunology Consultants Monkey IgG ELISA Kit
SDP
Supplier:  Immunology Consultants E85G
Encompass_Preferred
The Monkey IgG ELISA Kit is intended for the quantitative determination of total IgG in monkey biological samples.
Catalog No. 50-196-1941