Learn More
Kingfisher Biotech Inc Cynomolgus Monkey IL-1 beta Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1597Y025
Encompass
The Cynomolgus Monkey IL-1 beta yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey IL-1 beta applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey IL-1 beta yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey IL-1 beta Specifications: (Molecular Weight: 17.2 kDa) (Amino Acid Sequence: APVRSLHCTL RDAQLKSLVM SGPYELKALH LQGQDLEQQV VFSMSFVQGE ESNDKIPVAL GLKAKNLYLS CVLKDDKPTL QLESVDPKNY PKKKMEKRFV FNKIEINNKL EFESAQFPNW YISTSQAENM PVFLGGTRGG QDITDFTMQF VS (152)) (Gene ID: 102119749). For research use only. Made in the USA
Catalog No. 50-253-3118