Learn More
Invitrogen™ Monkey IL-10 ELISA Kit
Description
Includes
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey IL-10
Biotin-Conjugate anti-monkey IL-10 monoclonal antibody
Streptavidin-HRP
Monkey IL-10 Standard lyophilized, 500 U/ml upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
The Monkey Interleukin 10 (Mk IL-10) ELISA quantitates Mk IL-10 in monkey (baboon, cynomolgus) serum or cell culture medium. The assay will exclusively recognize both natural and recombinant monkey IL-10. Principle of the method The Monkey IL-10 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P79338 |
| Assay Range | 3.9-250 U/mL |
| Assay Sensitivity | 1.3 U/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Serum, Supernatant |
| Target Species | Baboon, Cynomolgus Monkey |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Show More |
Frequently Asked Questions (FAQs)
You can find expected "normal levels" listed in the Platinum ELISA Kit manual under "Expected Values". Please be aware that these normal values were obtained in our laboratories with a limited number of samples. We strongly suggest that each laboratory perform studies in order to establish normal range for their sample populations.
In case of small reagent volumes, please spin down vial before use to make sure to all contents are collected in the bottom of the tube.
We recommend following the instructions for sample dilutions as provided in the manual. We cannot guarantee optimal performance of the assay if internal assay dilutions as instructed were omitted.
Incubation times should be followed exactly as stated in the product manual to ensure optimal Platinum ELISA test performance.
An initial washing step is not necessary but recommended as accuracy may be increased.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.