Learn More
Invitrogen™ Monkey IL-15 ELISA Kit
Description
Monkey IL-15 quantitates monkey IL-15 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey IL-15.
Specifications
Specifications
| Accession Number | P40933 |
| Assay Range | 2.05-500 pg/mL |
| Assay Sensitivity | 2 pg/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human, Monkey |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15; |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.