Learn More
Kingfisher Biotech Inc Cynomolgus Monkey IL-2 Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1096Y005
Encompass
The Monkey IL-2 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Monkey IL-2 applications are for cell culture, ELISA standard, and Western Blot Control. Monkey IL-2 yeast-derived recombinant protein can be purchased in multiple sizes. Monkey IL-2 Specifications: (Molecular Weight: 15.5 kDa) (Amino Acid Sequence: APTSSSTKKT QLQLEHLLLD LQMILNGINN YKNPKLTRML TFKFYMPKKA TELRHLQCLE EELKPLEEVL NLAQSKSFHL RDTKDLISNI NVIVLELKGS ETTLMCEYAD ETATIVEFLN RWITFCQSII STLT (134)) (Gene ID: 102129830). For research use only. Made in the USA
Catalog No. 50-253-3673