Learn More
Abcam Monkey IL-2 ELISA Kit, 1 x 96 Tests
Catalog No. 502655198
Encompass_Preferred
Monkey IL-2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Monkey IL-2 with a sensitivity of 12 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-5198 Supplier Abcam Supplier No. AB2835551X96TESTS
Monkey IL-2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Monkey IL-2 with a sensitivity of 12 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement