Learn More
Abcam Monkey IL-2 ELISA Kit, 1 x 96 Tests
Catalog No. 502584769
Encompass_Preferred
Monkey IL-2 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Monkey IL-2 in Monkey in Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4769 Supplier Abcam Supplier No. AB1195411X96TESTS
Monkey IL-2 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Monkey IL-2 in Monkey in Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement