Learn More
Kingfisher Biotech Inc Cynomolgus Monkey IL-6 Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1017Y005
Encompass
The Cynomolgus Monkey IL-6 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey IL-6 applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey IL-6 yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey IL-6 Specifications: (Molecular Weight: 21.0 kDa) (Amino Acid Sequence: APVLPGEDSK DVAAPHSQPL TSSERIDKHI RYILDGISAL RKETCNRSNM CESSKEALAE NNLNLPKMAE KDGCFQSGFN EDTCLVKIIT GLLEFEVYLE YLQNRFESSE EQARAVQMST KVLIQFLQKK AKNLDAITTP EPTTNASLLT KLQAQNQWLQ DMTTHLILRS FKEFLQSSLR ALRQM (185)) (Gene ID: 102138971). For research use only. Made in the USA
Catalog No. 50-241-4044