Learn More
Kingfisher Biotech Inc Rabbit CCL21 ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1169Y003
Encompass_Preferred
The Rabbit CCL21 (6Ckine) ELISA contains capture antibody, standard, and detection antibody for development of a Rabbit CCL21 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include 6Ckine, CKb9, ECL, SCYA21, SLC, TCA4. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5351