Learn More
American Research Products Inc Monkey Interleukin 1 alpha IL
Catalog No. 5000326106
Encompass
Monkey Interleukin-1 alpha (IL1A) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26106 Supplier American Research Products Inc Supplier No. CSBE10041MO
Monkey Interleukin-1 alpha (IL1A) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich