Learn More
American Research Products Inc Monkey Interleukin 12 IL 12 P
Catalog No. 5000326728
Encompass
Monkey Interleukin 12 IL-12/P70 ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-26728 Supplier American Research Products Inc Supplier No. CSBE08904MK
Monkey Interleukin 12 IL-12/P70 ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma Method Sandwich