Learn More
American Research Products Inc Monkey Interleukin 15 IL 15 E
Catalog No. 5000328856
Encompass
Monkey Interleukin 15 IL-15 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-28856 Supplier American Research Products Inc Supplier No. CSBE14392MK
Monkey Interleukin 15 IL-15 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma Method Sandwich