Learn More
American Research Products Inc Monkey Interleukin 17A IL 17A
Catalog No. 5000328788
Encompass
Monkey Interleukin 17A (IL-17A/IL-17) ELISA Kit from CUSABIO Detection Range 9 38 pg/mL-600 pg/mL Sensitivity 4 69 pg/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-28788 Supplier American Research Products Inc Supplier No. CSBE08905MK
Monkey Interleukin 17A (IL-17A/IL-17) ELISA Kit from CUSABIO Detection Range 9 38 pg/mL-600 pg/mL Sensitivity 4 69 pg/mL Sample serum plasma cell culture supernates Method Sandwich