Learn More
American Research Products Inc Monkey Interleukin 18 IL18 E
Catalog No. 5000329230
Encompass
Monkey Interleukin-18 (IL18) ELISA Kit from CUSABIO Detection Range 47 pg/mL-3000 pg/mL Sensitivity 23 5 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29230 Supplier American Research Products Inc Supplier No. CSBE08979MK
Monkey Interleukin-18 (IL18) ELISA Kit from CUSABIO Detection Range 47 pg/mL-3000 pg/mL Sensitivity 23 5 pg/mL Sample serum plasma Method Sandwich