Learn More
American Research Products Inc Monkey Interleukin 6 IL 6 ELI
Supplier:  American Research Products Inc CSBE10050MO
Encompass
Monkey Interleukin 6 IL-6 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29078