Learn More
American Research Products Inc Monkey Interleukin 6 IL 6 ELI
Catalog No. 5000329171
Encompass
Monkey Interleukin 6 IL-6 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 15 625 pg/mL Sample cell culture supernates Method Sandwich
Catalog No. 50-003-29171 Supplier American Research Products Inc Supplier No. CSBE10050MOIS
Monkey Interleukin 6 IL-6 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 15 625 pg/mL Sample cell culture supernates Method Sandwich