Learn More
American Research Products Inc Monkey Interleukin 8 IL 8 ELI
Catalog No. 5000326498
Encompass
Monkey Interleukin-8 IL-8 ELISA Kit from CUSABIO Detection Range 7 8pg/mL-500pg/mL Sensitivity 3 9pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26498 Supplier American Research Products Inc Supplier No. CSBE08051MOIS
Monkey Interleukin-8 IL-8 ELISA Kit from CUSABIO Detection Range 7 8pg/mL-500pg/mL Sensitivity 3 9pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich