Learn More
American Research Products Inc Monkey Interleukin 8 IL 8 ELI
Supplier:  American Research Products Inc CSBE08051MO
Encompass
Monkey Interleukin-8 IL-8 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26507