Learn More
American Research Products Inc Monkey Macrophage Inflammatory
Catalog No. 5000328154
Encompass
Monkey Macrophage Inflammatory Protein-3a (MIP-3a/CCL20) ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28154 Supplier American Research Products Inc Supplier No. CSBE17925MK
Monkey Macrophage Inflammatory Protein-3a (MIP-3a/CCL20) ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich