Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Monkey PF-4 ELISA Kit

Catalog No. EP17RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EP17RB 96 Tests
EP17RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EP17RB Supplier Invitrogen™ Supplier No. EP17RB
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Monkey PF-4 quantitates monkey PF-4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey PF-4.

Platelet factor-4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair (Eisman et al., 1990).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; CXCL4; Endothelial cell growth inhibitor; Iroplact; Oncostatin-A; Pf4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4
Assay Range 0.041-10 ng/mL
Assay Sensitivity 0.041 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Monkey
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; Cxcl4; Endothelial cell growth inhibitor; Iroplact; oncostatin-A; PF4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; Cxcl4; Endothelial cell growth inhibitor; Iroplact; oncostatin-A; PF4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4;
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target PF4
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.