Learn More
American Research Products Inc Monkey Progesterone PROG ELI
Catalog No. 5000326051
Encompass
Monkey Progesterone (PROG) ELISA Kit from CUSABIO Detection Range 1 ng/ml-70 ng/ml Sensitivity 0 2 ng/mL Sample serum plasma Method Competitive
Catalog No. 50-003-26051 Supplier American Research Products Inc Supplier No. CSBE13352MK
Monkey Progesterone (PROG) ELISA Kit from CUSABIO Detection Range 1 ng/ml-70 ng/ml Sensitivity 0 2 ng/mL Sample serum plasma Method Competitive