Learn More
American Research Products Inc Monkey serum amyloid A1 SAA1
Catalog No. 5000328202
Encompass
Monkey serum amyloid A1 (SAA1) ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28202 Supplier American Research Products Inc Supplier No. CSBEL020656RH
Monkey serum amyloid A1 (SAA1) ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Competitive