Learn More
American Research Products Inc Monkey thyroid stimulating hor
Catalog No. 5000327963
Encompass
Monkey thyroid stimulating hormone (TSH) ELISA kit from CUSABIO Detection Range 0 3 IU/mL-12 IU/mL Sensitivity 0 15 IU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27963 Supplier American Research Products Inc Supplier No. CSBEQ027364MK
Monkey thyroid stimulating hormone (TSH) ELISA kit from CUSABIO Detection Range 0 3 IU/mL-12 IU/mL Sensitivity 0 15 IU/mL Sample serum plasma tissue homogenates Method Sandwich