Learn More
American Research Products Inc Monkey tri iodothyronine T3
Catalog No. 5000327406
Encompass
Monkey tri-iodothyronine (T3) ELISA kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27406 Supplier American Research Products Inc Supplier No. CSBEQ027510MK
Monkey tri-iodothyronine (T3) ELISA kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma tissue homogenates Method Competitive