Learn More
American Research Products Inc Monkey Tumor necrosis factor a
Catalog No. 5000330095
Encompass
Monkey Tumor necrosis factor a TNF-a ELISA KIT from CUSABIO Detection Range 4 69 pg/mL-300 pg/mL Sensitivity 2 345 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30095 Supplier American Research Products Inc Supplier No. CSBE10039MO
Monkey Tumor necrosis factor a TNF-a ELISA KIT from CUSABIO Detection Range 4 69 pg/mL-300 pg/mL Sensitivity 2 345 pg/mL Sample serum plasma tissue homogenates Method Sandwich