Learn More
American Research Products Inc Monkey Vascular Endothelial ce
Catalog No. 5000326804
Encompass
Monkey Vascular Endothelial cell Growth Factor (VEGF) ELISA Kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma cell culture supernates cell lysates Method Sandwich
Catalog No. 50-003-26804 Supplier American Research Products Inc Supplier No. CSBE16502MK
Monkey Vascular Endothelial cell Growth Factor (VEGF) ELISA Kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma cell culture supernates cell lysates Method Sandwich