Learn More
American Research Products Inc Monkey Vitamin K dependent pro
Catalog No. 5000327383
Encompass
Monkey Vitamin K-dependent protein S (PROS1) ELISA kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27383 Supplier American Research Products Inc Supplier No. CSBEL018754RH
Monkey Vitamin K-dependent protein S (PROS1) ELISA kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma tissue homogenates Method Competitive