Learn More
Genscript Corporation Monkeypox A29L protein His Ta
Catalog No. 5000056677
Encompass
The A29L protein from Monkeypox binds to cell surface heparin to promote fusion of viral membrane with host plasma membrane it is a potential target for developing diagnosis and vaccine
Catalog No. 50-000-56677 Supplier Genscript Corporation Supplier No. Z037521
Add to Cart
Add to Cart
The A29L protein from Monkeypox binds to cell surface heparin to promote fusion of viral membrane with host plasma membrane it is a potential target for developing diagnosis and vaccine